missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDC2/CDK1 Monoclonal antibody specifically detects CDC2/CDK1 in S. pombe samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CDC2/CDK1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | Y100.4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000 |
| Formulation | PBS |
| Gene Alias | CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?