missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC2/CDK1 Antibody (Y100.4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £386.00
Specifications
| Antigen | CDC2/CDK1 |
|---|---|
| Clone | Y100.4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18481109
|
Novus Biologicals
NB100-2716-0.025mg |
0.025 mg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18425319
|
Novus Biologicals
NB100-2716 |
0.1 mg |
£386.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC2/CDK1 Monoclonal antibody specifically detects CDC2/CDK1 in S. pombe samples. It is validated for Western BlotSpecifications
| CDC2/CDK1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| PBS | |
| 983 | |
| IgG2b | |
| Protein A purified |
| Y100.4 | |
| Western Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| S. pombe | |
| CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2 | |
| Peptide spanning the C-terminal 14 residues of S. pombe CDC2/CDK1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title