missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC16, p Thr580 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-889
This item is not returnable.
View return policy
Description
CDC16 Polyclonal specifically detects CDC16 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CDC16 (p Thr580) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| ANAPC6subunit 6, CDC16 (cell division cycle 16, S. cerevisiae, homolog), CDC16 cell division cycle 16 homolog (S. cerevisiae), CDC16 homolog, CDC16Hs, cell division cycle 16 homolog (S. cerevisiae), cell division cycle protein 16 homolog, Cyclosome subunit 6 | |
| Rabbit | |
| 71.65 kDa | |
| 0.1 mg | |
| Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway | |
| 8881 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 0.97 mg/ml | |
| Western Blot 1:500-1:3000, ELISA 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Q13042 | |
| CDC16 | |
| This affinity purified CDC16 [p Thr580] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region surrounding pT580 of Human CDC16 protein. (Uniprot: Q13042) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This product is an affinity purified antibody produced by immunoaffinity chromatography using phospho peptide coupled to agarose beads followed by solid phase adsorption(s) against non-phospho peptide and non-specific peptide to remove any unwanted reactivities. This antibody is specific for phosphorylated human CDC16 protein at the pT580 residue. BLAST analysis indicates 100 % homology of the immunizing sequence with CDC16 from human. No cross reactivity is expected against homologues from other sources. This antibody is expected to react with all isoforms of this protein. Minimal reactivity is expected with the non-phosphorylated form of the protein. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction