missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDC16 Monoclonal antibody specifically detects CDC16 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | CDC16 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol, 0.05% BSA |
| Gene Alias | ANAPC6subunit 6, CDC16 (cell division cycle 16, S. cerevisiae, homolog), CDC16 cell division cycle 16 homolog (S. cerevisiae), CDC16 homolog, CDC16Hs, cell division cycle 16 homolog (S. cerevisiae), cell division cycle protein 16 homolog, Cyclosome subunit 6 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200-300 of human CDC16 (Q13042).,, Sequence:, LLRFLFENKLKKYNKPSETVIPESVDGLQENLDVVVSLAERHYYNCDFKMCYKLTSVVMEKDPFHASCLPVHIGTLVELNKANELFYLSHKLVDLYPSNPV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?