Learn More
Invitrogen™ Cdc14A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114232
Description
Immunogen sequence: DDDVEMKNGI TQGDKLRALK SQRQPRTSPS CAFRSDDTKG HPRAVSQPFR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 87%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%.
The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms.
Specifications
| Cdc14A | |
| Polyclonal | |
| Unconjugated | |
| CDC14A | |
| A830059A17Rik; CDC10 (cell division cycle 10, S. cerevisiae, homolog); cdc14; CDC14 cell division cycle 14 homolog A; CDC14 cell division cycle 14A; CDC14A; CDC14a1; CDC14A2; cell division cycle 14A; DFNB105; Dual specificity protein phosphatase CDC14A; hCDC14 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8556 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UNH5 | |
| CDC14A | |
| Immunogen sequence: DDDVEMKNGI TQGDKLRALK SQRQPRTSPS CAFRSDDTKG HPRAVSQPFR (aa 384-433). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.