missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDC123 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Gene ID (Entrez) | 8872 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 27 kDa |
| Quantity | 100 μL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | cell division cycle 123 homolog (S. cerevisiae), cell division cycle protein 123 homolog, chromosome 10 open reading frame 7, D123C10orf7, FLJ13863, HT-1080, Protein D123, PZ32 |
| Gene Symbol | CDC123 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-68733. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL |
| Research Category | Cell Cycle and Replication |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction