missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDAN1 Antibody (Cod177), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50176CL1
This item is not returnable.
View return policy
Description
CDAN1 Monoclonal antibody specifically detects CDAN1 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CDAN1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CDA1, CDAI, CDA-I, codanin, codanin-1, congenital dyserythropoietic anemia, type I, discs lost homolog, DLT, PRO1295 | |
| aa 294-607 of cloned human protein | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 146059 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Cod177 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction