missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDA Monoclonal Antibody (K1E006_4H7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555855
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: MAQKRPACTL KPECVQQLLV CSQEAKKSAY CPYSHFPVGA ALLTQEGRIF KGCNIENACY PLGICAERTA IQKAVSEGYK DFRAIAIASD MQDDFISPCG ACRQVMREFG TNWPVYMTKP DGTYIVMTVQ ELLPSSFGPE DLQKTQ
This gene encodes an enzyme involved in pyrimidine salvaging. The encoded protein forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. It is one of several deaminases responsible for maintaining the cellular pyrimidine pool. Mutations in this gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias.
Specifications
| CDA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P32320 | |
| CDA | |
| Recombinant human CDA (immunogen range is 1-146). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K1E006_4H7 | |
| Unconjugated | |
| CDA | |
| 2210401N16Rik; CDA; CDD; cytidine aminohydrolase; Cytidine deaminase; cytosine nucleoside deaminase; EC 3.5.4.5; small cytidine deaminase | |
| Mouse | |
| Protein A | |
| RUO | |
| 978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction