missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDA Monoclonal Antibody (K1E006_15H12), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA555962

 View more versions of this product

Product Code. 30205507

  • £453.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: MAQKRPACTL KPECVQQLLV CSQEAKKSAY CPYSHFPVGA ALLTQEGRIF KGCNIENACY PLGICAERTA IQKAVSEGYK DFRAIAIASD MQDDFISPCG ACRQVMREFG TNWPVYMTKP DGTYIVMTVQ ELLPSSFGPE DLQKTQ

This gene encodes an enzyme involved in pyrimidine salvaging. The encoded protein forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. It is one of several deaminases responsible for maintaining the cellular pyrimidine pool. Mutations in this gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDA
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
P32320
CDA
Recombinant human CDA (immunogen range is 1-146).
50 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
K1E006_15H12
Unconjugated
CDA
2210401N16Rik; CDA; CDD; cytidine aminohydrolase; Cytidine deaminase; cytosine nucleoside deaminase; EC 3.5.4.5; small cytidine deaminase
Mouse
Protein A
RUO
978
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.