missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD99L2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA548067
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD99-L2 is observed and less than 5% cross-reactivity with recombinant mouse CD99 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD99L2 may function as a homophilic adhesion molecule. CD99L2 functions in leukocyte-endothelial cell interactions during leukocyte extravasation, and in particular, at the diapedesis step. CD99L2 does not seem to be involved in docking of leukocytes to the vessel wall or in lymphocyte diapedesis.
Specifications
| CD99L2 | |
| Polyclonal | |
| Unconjugated | |
| Cd99l2 | |
| AW548191; CD99; Cd99 antigen-like 2; CD99 antigen-like protein 2; CD99 molecule like 2; CD99 molecule-like 2; CD99B; CD99L2; MIC2 (monoclonal Imperial Cancer Research Fund 2)-like 1; MIC2 like 1; MIC2 like-1; MIC2L1; MIC2-like protein 1; protein rhombex-40; Rhombencephalic expression protein 40 kDa; Rhombex40; Rhombex-40; single-pass transmembrane protein; UNQ1964/PRO4486; vms-tm2; Xap89 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 171486 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8BIF0 | |
| Cd99l2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD99-L2 Val20-Ala141. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction