missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Rabbit anti-Human, Clone: MIC2/3478R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07315-100ug
This item is not returnable.
View return policy
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| CD99 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| Synthetic peptide corresponding to CD99 residues within aa85 C terminal of CD99 was used as an immunogen. | |
| 100 μg | |
| Immunology | |
| 4267 | |
| Store at 4°C. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| MIC2/3478R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction