Learn More
Invitrogen™ CD99 Monoclonal Antibody (3B2/TA8), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528580
Description
This antibody recognizes an extracellular epitope of CD99, an approximately 32 kDa sialoglycoprotein expressed on the surface of many cell types, with particularly strong expression on Ewing's sarcoma and peripheral primitive neuroectodermal tumors. Within the hematopoietic system, CD99 is expressed on virtually all cell types except granulocytes. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.
Specifications
| CD99 | |
| Monoclonal | |
| APC | |
| CD99 | |
| 1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 4267 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 3B2/TA8 | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| P14209 | |
| CD99 | |
| Human thymocytes. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.