Learn More
CD99/MIC2 (Ewing' s Sarcoma Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Detect CD99 in human and rat samples with Epredia™ CD99/MIC2 (Ewing's Sarcoma Marker) Ab-1, Mouse Monoclonal Antibody, which provides accurate, reproducible results.
Brand: Epredia MS-294-R7
Description
CD99, or MIC2 gene product, or E2 antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. The antigen is also expressed by most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewing's sarcoma cells and primitive peripheral neuroectodermal tumors.Host Species: Mouse
Clone: HO36-1.1
Isotype: IgM
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified E-rosette forming cells from human peripheral blood lymphocytes
Molecular Weight: 27-32kDa
Positive Control: Pancreas or Ewing's sarcoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
| CD99/MIC2 (Ewings Sarcoma Marker) Ab-1 | |
| Monoclonal | |
| — | |
| Mouse | |
| 7 mL | |
| Oncogenesis | |
| Human, Rat | |
| IgM |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HO36-1.1 | |
| Unconjugated | |
| Purified E-rosette forming cells from human peripheral blood lymphocytes | |
| IVD | |
| Primary | |
| Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.