missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99-L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98286-100ul
This item is not returnable.
View return policy
Description
CD99-L2 Polyclonal antibody specifically detects CD99-L2 in Mouse samples. It is validated for ELISA
Specifications
| CD99-L2 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| CD99 antigen, CD99 antigen-like 2, CD99 antigen-like protein 2, CD99 molecule-like 2, CD99B, DKFZp761H2024, MIC2 like 1, MIC2L1, MIC2-like protein 1 | |
| Produced in rabbits immunized with purified, recombinant Mouse CD99-L2 (Accession#: NP_612182.1; Met 1-Ala 164) | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 83692 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction