missing translation for 'onlineSavingsMsg'
Learn More

CD99-L2 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30736538 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736538 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736538 Supplier Novus Biologicals Supplier No. AF5185JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD99-L2 Polyclonal antibody specifically detects CD99-L2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD99-L2
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias CD99 antigen, CD99 antigen-like 2, CD99 antigen-like protein 2, CD99 molecule-like 2, CD99B, DKFZp761H2024, MIC2 like 1, MIC2L1, MIC2-like protein 1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD99-L2, Val20-Ala188, Accession # Q8TCZ2
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83692
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.