missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Antibody (OTI1D5), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71197MFV610
This item is not returnable.
View return policy
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Flow Cytometry
Specifications
| CD99 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| OTI1D5 | |
| Flow Cytometry | |
| 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| Full length human recombinant protein of human CD99 (NP_002405) produced in HEK293T cell. | |
| 0.1 mL | |
| Immunology | |
| 4267 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction