missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Antibody (MEM-131), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-41282
This item is not returnable.
View return policy
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| CD99 | |
| Monoclonal | |
| PE | |
| P14209 | |
| CD99 | |
| HPB-ALL human peripheral blood leukemia T-cell line | |
| 100 Tests | |
| Immunology | |
| 4267 | |
| Human | |
| IgM |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| MEM-131 | |
| Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody MEM-131 reacts with CD99R, an epitope restricted to a subset of CD99 molecule expressed on myeloid cells, NK cells and T lymphocytes. HLDA V; WS Code AS S020 HLDA V; WS Code T T-E2.02 HLDA V; WS Code T T-017 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction