missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Antibody (DN16), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64930PEATT594
This item is not returnable.
View return policy
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD99 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| CD99 | |
| Made to Human CD99 | |
| 0.1 mL | |
| Primary | |
| Recognizes the human CD99 cell surface antigen, a 32kD glycoprotein expressed by all leucocytes. CD99 expression tends to be highest on immature cells. Functionally CD99 has been found to be involved in lymphocyte adhesion/aggregation and apoptosis. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| DN16 | |
| Flow Cytometry | |
| 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
| Mouse | |
| Protein A or G purified | |
| Immunology | |
| 4267.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction