Learn More
CD99 Antibody (CL12929) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18550FR
Product Code. 30297599
  • £582.00 / 0.10mL
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD99
Monoclonal
DyLight 680
12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
This antibody was developed using a synthetic peptide derived from P14209, with the exact immunogen sequence remaining proprietary.
0.1 mL
Immunology
4267
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12929
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified