missing translation for 'onlineSavingsMsg'
Learn More

CD98hc Monoclonal Antibody (5E5), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£69.50 - £203.00

Specifications

Antigen CD98hc
Clone 5E5
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15536466
View Documents
Invitrogen
11-0982-41
25 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15546466
GREENER_CHOICE
View Documents
Invitrogen™
11-0982-42
100 Tests
£203.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The monoclonal antibody 5E5 recognizes human CD98hc protein also called CD98 heavy chain, FRP-1 (fusion regulatory protein-1 ) or 4F2. CD98 is a disulfide-linked complex composed of a heavy chain (a Type II membrane protein) and a light chain of which there are more than six different chains (LAT1, LAT2, y+LAT1, y+LAT2, Asc-1, xCT). CD98 can associate directly or indirectly with beta 1 integrins, EpCAM and CD147 (EMMPRIN). The function of CD98 is thought to involve amino acid transport, cell fusion concomitantly with increased plasma membrane expression and integrin mediated adhesion. Expression of CD98 is broad including non-hematopoietic cells. In peripheral blood cells, monocytes expression is high while lymphocytes have intermediate levels and granulocytes express very little. Applications Reported: This 5E5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 5E5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD98 (4F2) is a type II transmembrane glycoprotein which serves as the heavy chain of the heterodimeric amino acid transporters (HATs). CD98, linked to various light chains by disulfide bond, is responsible for cell surface expression and basolateral localization of this transporter complex in polarized epithelial cells and also interacts with beta1 integrins and increases their affinity for ligand. Besides its roles in amino acid transport, CD98 is thus involved in cell fusion and activation. It is implicated in regulation of cellular differentiation, growth and apoptosis.
Specifications

Specifications

CD98hc
5 μL/Test
Monoclonal
Liquid
RUO
P08195
SLC3A2
Primary
4° C, store in dark, DO NOT FREEZE!
SLC3A2
5E5
Flow Cytometry
FITC
Mouse
Human
6520
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.