Learn More
Invitrogen™ CD98 Recombinant Rabbit Monoclonal Antibody (071)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA529574
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CD98/SLC3A2.
CD98 (4F2) is a type II transmembrane glycoprotein which serves as the heavy chain of the heterodimeric amino acid transporters (HATs). CD98, linked to various light chains by disulfide bond, is responsible for cell surface expression and basolateral localization of this transporter complex in polarized epithelial cells and also interacts with beta1 integrins and increases their affinity for ligand. Besides its roles in amino acid transport, CD98 is thus involved in cell fusion and activation. It is implicated in regulation of cellular differentiation, growth and apoptosis.
Specifications
| CD98 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P08195 | |
| SLC3A2 | |
| Recombinant Human CD98/SLC3A2 Protein (Arg206-Ala630). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 071 | |
| Unconjugated | |
| SLC3A2 | |
| 4F2; 4F2 cell-surface antigen heavy chain; 4F2 heavy chain antigen; 4F2hc; 4T2HC; AI314110; antigen defined by monoclonal antibody 4F2, heavy chain; antigen identified by monoclonal antibodies 4F2; antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43; Cd98; CD98 antigen; CD98 heavy chain; CD98HC; Ly10; Ly-10; Ly-m10; Lymphocyte activation antigen 4F2 large subunit; MDU1; Mgp-2hc; monoclonal antibody 44D7; NACAE; RGD:3073}; SLC3A2; slc3a2 {ECO:0000312; solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2; solute carrier family 3 (amino acid transporter heavy chain), member 2; solute carrier family 3 member 2; solute carrier family 3, member 2; type II transmembrane protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.