missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD98 Antibody (1C11.7E3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£183.00 - £339.00
Specifications
| Antigen | CD98 |
|---|---|
| Clone | 1C11.7E3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug ml, Flow Cytometry 2.5 ug million cells, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Flow (Cell Surface) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18110459
|
Novus Biologicals
NBP2-36491SS |
0.025 mg |
£183.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18796813
|
Novus Biologicals
NBP2-36491 |
0.1 mg |
£339.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD98 Monoclonal specifically detects CD98 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface).Specifications
| CD98 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen | |
| SLC3A2 | |
| IgG2b κ | |
| Protein G purified |
| 1C11.7E3 | |
| Western Blot 2 ug ml, Flow Cytometry 2.5 ug million cells, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Flow (Cell Surface) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P08195 | |
| 6520 | |
| Partial recominant human CD98 (between amino acids 200-400) [UniProt P08195] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title