missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13071-1-AP-150UL
This item is not returnable.
View return policy
Description
CD97 is a G-protein-coupled seven-span transmembrane adhesive receptor that is constitutively expressed on granulocytes and monocytes and rapidly upregulated on T and B cells upon activation. CD97 is produced in alternatively spliced forms and its cellular ligand is CD55 (DAF), which protects various cell types from complement-mediated damage. Interaction of CD97 on leukocytes and CD55 on vessel cells probably facilitate leukocyte activation and migration into the tissues, similarly, CD97 seems to play a role in tumour migration and invasiveness. CD97 is involved in T cell regulation and peripheral granulocyte homeostasis.Specifications
| CD97 | |
| Polyclonal | |
| Unconjugated | |
| CD97 | |
| CD97, CD97 antigen, CD97 molecule, Leukocyte antigen CD97, TM7LN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 976 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48960 | |
| CD97 | |
| CD97 Fusion Protein Ag3745 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction