Learn More
CD97 Isoform 1 Antibody (380903), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2529MFV450
394.40 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18525938
  • £494.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
CD97 Isoform 1 Monoclonal antibody specifically detects CD97 Isoform 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD97 Isoform 1
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
976
Store at 4C in the dark.
IgG2a
Western Blot, Flow Cytometry, Neutralization, CyTOF
380903
Western Blot, Flow Cytometry, Neutralization, CyTOF-ready
CD97, TM7LN1
Mouse myeloma cell line NS0-derived recombinant human CD97, Gln21-Gln398, Accession # NP_001775.2
0.1 mL
Primary
Human
Purified