missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD97 Polyclonal antibody specifically detects CD97 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Applications | Western Blot, Immunocytochemistry, Immunoprecipitation |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Host Species | Rabbit |
| Primary or Secondary | Primary |
| Form | Purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?