missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Antibody (MEM-180), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-425AF532
This item is not returnable.
View return policy
Description
CD97 Monoclonal specifically detects CD97 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD97 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ADGRE5 | |
| PHA-activated peripheral blood cells | |
| Primary | |
| The antibody MEM-180 recognizes an unique epitope on CD97, a 75-85 kDa surface glycoprotein of G-protein-coupled receptor family, expressed on activated B and T lymphocytes, monocytes/macrophages, dendritic cells and granulocytes. HLDA VI; WS Code BP 415 HLDA VI; WS Code NL N-L023 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| MEM-180 | |
| Flow Cytometry | |
| CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
| Mouse | |
| 0.1 mL | |
| 976 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction