missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD97 Polyclonal specifically detects CD97 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | CD97 |
| Applications | ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | PBS |
| Gene Alias | CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant CD97 (NP_001775.2; Met1-Gln398). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?