missing translation for 'onlineSavingsMsg'
Få mere at vide

CD97 Antibody, DyLight 405, Novus Biologicals™

Artikelnummer. 18349876 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
Pakningsstørrelse:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18349876 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18349876 Leverandør Novus Biologicals Leverandørnr. NBP299765V

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

CD97 Polyclonal specifically detects CD97 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CD97
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CD97 (NP_001775.2; Met1-Gln398).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 976
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.