missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD96 Rabbit anti-Mouse, Clone: 6A6, Novus Biologicals™
Rabbit Monoclonal Antibody
£200.00 - £444.00
Specifications
| Antigen | CD96 |
|---|---|
| Clone | 6A6 |
| Dilution | Flow Cytometry, ELISA, Block/Neutralize |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18361403
|
Novus Biologicals
NBP3-12037-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18344242
|
Novus Biologicals
NBP3-12037-0.2MG |
0.2 mg |
£444.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD96 Monoclonal antibody specifically detects CD96 in Mouse samples. It is validated for Flow Cytometry, ELISA, NeutralizationSpecifications
| CD96 | |
| Flow Cytometry, ELISA, Block/Neutralize | |
| Unconjugated | |
| RUO | |
| PBS | |
| 10225 | |
| IgG λ | |
| Protein A purified |
| 6A6 | |
| Monoclonal | |
| Rabbit | |
| Mouse | |
| CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile | |
| This antibody was raised by immunising rats with murine CD96. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title