missing translation for 'onlineSavingsMsg'
Learn More

CD96 Antibody (6A6), DyLight 350, Novus Biologicals™

Product Code. 18381082 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381082 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381082 Supplier Novus Biologicals Supplier No. NBP312037UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD96 Monoclonal antibody specifically detects CD96 in Mouse samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD96
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 6A6
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile
Host Species Rabbit
Immunogen This antibody was raised by immunising rats with murine CD96.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10225
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.