missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD96 Antibody (628211) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6199JF669
This item is not returnable.
View return policy
Description
CD96 Monoclonal antibody specifically detects CD96 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CD96 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile | |
| Mouse myeloma cell line NS0-derived recombinant human CD96, Val22-Asp501, Accession # NP_005807 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 628211 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 10225 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction