Learn More
CD95 Mouse anti-Human, PE, Clone: DX2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555674
Description
Reacts with the 45kDa transmembrane cell surface Fas or Apo-1 antigen (designated CD95 at the Fifth HLDA Workshop) which is expressed on a variety of normal and neoplastic cells. The Fas/Apo-1 antigen is a polypeptide, which plays a role in the programmed sequence of events leading to cell death, termed apoptosis. The DX2 clone specifically reacts with murine L cells, murine L1210 leukemia cells and murine P815 mastocytoma cells transfected with human Fas cDNA but not with untransfected parental cell lines. Cross-linking with DX2 delivers an apoptotic signal indicating that DX2 recognizes a functional epitope of the CD95 antigen. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: DX2
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD95 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CD95-transfected L Cells | |
| 100 Tests | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Human, Pig, Rhesus Monkey | |
| IgG1 κ |
| Flow Cytometry | |
| DX2 | |
| APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.