Learn More
CD95 Mouse anti-Human, PE, Clone: DX2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556641
Description
Reacts with the human form of the 40-50kDa transmembrane cell surface Fas or Apo-1 antigen. The Fas/Apo-1 antigen is a polypeptide which plays a role in the programmed sequence of events leading to cell death, termed apoptosis. DX2 cross-reacts with a subset of peripheral blood lymphocytes, monocytes and granulocytes of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on lymphocytes is similar to that seen with human peripheral blood lymphocytes being broadly expressed on all major lymphocyte subsets. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: DX2
Isotype: IgG1, κ
Cross Reactivity: Baboon, Cynomolgus
Flow Cytometry
Specifications
| CD95 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CD95-transfected L Cells | |
| 50 Tests | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Human, Pig, Rhesus Monkey | |
| IgG1 κ |
| Flow Cytometry | |
| DX2 | |
| APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.