missing translation for 'onlineSavingsMsg'
Learn More

CD95 (Fas) Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BB700, Clone: DX2, BD Horizon™

Mouse Monoclonal Antibody

£166.06 - £396.33

Specifications

Antigen CD95 (Fas)
Clone DX2
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Blue 700 (BB700)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17207504
View Documents
BD Biosciences
566543
25 Tests
ONLINE EXCLUSIVE PRICE
£174.00
£166.06 / 25 tests
Save £7.94
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17297494
View Documents
BD Biosciences
566542
100 Tests
ONLINE EXCLUSIVE PRICE
£414.00
£396.33 / 100 tests
Save £17.67
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD95 (Fas) Monoclonal antibody specifically detects CD95 (Fas) in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD95 (Fas)
Flow Cytometry
Brilliant Blue 700 (BB700)
RUO
IgG1 κ
Refrigerate (2 to 8°C)
DX2
Monoclonal
Mouse
Human, Baboon, Cynomolgus, Rhesus Monkey
Primary
CD95 (Fas)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.