missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD93 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552664
This item is not returnable.
View return policy
Description
Immunogen sequence: FNTQGSFHCG CLPGWVLAPN GVSCTMGPVS LGPPSGPPDE EDKGEKEGST VPRAATASPT RGPEGTPKAT PTTSRPSLSS DAPITSAPLK MLAPSGSSGV WREPSIHHAT AASGPQEPAG GDSSVATQNN DGTDGQKL Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 29%.
The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton.
Specifications
| CD93 | |
| Polyclonal | |
| Unconjugated | |
| Cd93 | |
| 6030404G09Rik; AA145088; Aa4; AA4.1; AW555904; C1q receptor; C1q receptor 1; C1q/MBL/SPA receptor; C1qR; C1qR(P); C1qr1; C1qRP; Cd93; CD93 antigen; CD93 molecule; CDw93; Cell surface antigen AA4; complement component 1 q subcomponent receptor 1; complement component 1, q subcomponent, receptor 1; complement component C1q receptor; dJ737E23.1; ECSM3; Ly68; ly-68; Lymphocyte antigen 68; matrix-remodeling-associated protein 4; matrix-remodelling associated 4; MXRA4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22918 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPY3 | |
| Cd93 | |
| Recombinant protein corresponding to Human CD93. Recombinant protein control fragment (Product #RP-90184). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction