missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD93, Mouse anti-Human, Clone: 3D12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00022918-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: GTACYTAHSGKLSAAEAQNHCNQNGGNLATVKSKEEAQHVQRVLAQLLRREAALTARMSKFWIGLQREKGKCLDPSLPLKGFSWVGGGEDTPYSNWHKELRNSCISKRSpecifications
| CD93 | |
| Monoclonal | |
| Unconjugated | |
| CD93 molecule | |
| C1QR1/C1qR(P)/C1qRP/CDw93/MXRA4/dJ737E23.1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3D12 | |
| In 1x PBS, pH 7.4 | |
| NM_012072 | |
| CD93 | |
| CD93 (NP_036204, 33 a.a. ∼ 140 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 22918 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction