missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD91 Monoclonal Antibody (A2MR-a2), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£97.75 - £268.00
Specifications
Antigen | CD91 |
---|---|
Clone | A2MR-a2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-eFluor 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15509166
|
Affymetrix eBioscience
46-0919-42 |
100 Tests |
£268.00
100 tests |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15599156
|
Affymetrix eBioscience
46-0919-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The protein encoded by this gene is an endocytic receptor involved in several cellular processes, including intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. In addition, the encoded protein is necessary for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of this gene decreases with age and has been found to be lower than controls in brain tissue from Alzheimer patients.
CD91 Monoclonal antibody specifically detects CD91 in Human samples. It is validated for Flow Cytometry.Specifications
CD91 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
4035 | |
IgG1 κ | |
Affinity chromatography | |
LRP1 |
A2MR-a2 | |
Monoclonal | |
Liquid | |
RUO | |
Q07954 | |
LRP1 | |
Primary | |
Antibody |
For Research Use Only.