missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD91 Monoclonal Antibody (A2MR-a2), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £340.00
Specifications
| Antigen | CD91 |
|---|---|
| Clone | A2MR-a2 |
| Concentration | 0.05 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056098
|
Invitrogen™
H073T03R04-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055780
|
Invitrogen™
H073T02R04-A |
25 Tests |
£154.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD91 Monoclonal antibody specifically detects CD91 in Human samples. It is validated for Flow CytometrySpecifications
| CD91 | |
| 0.05 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q07954 | |
| 4035 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| A2MR-a2 | |
| Flow Cytometry | |
| NovaFluor Red 710 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A2MR; AI316852; alpha 2-macroglobulin receptor; alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; APR; b2b1554Clo; CD91; FLJ16451; IGFBP3R; LDL receptor related protein 1; lipoprotein receptor-related protein; low density lipoprotein receptor-related protein 1; low density lipoprotein-related protein 1 (alpha-2-macroglobulin receptor); Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; Lrp; Lrp1; LRP-1; LRP1A; LRP-515; LRP-85; LRPICD; MGC88725; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; TGFBR5; type V tgf-beta receptor | |
| IgG1 κ | |
| Affinity chromatography | |
| LRP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title