missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD90 (Thy1) Recombinant Rabbit Monoclonal Antibody (2V9S6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542657
This item is not returnable.
View return policy
Description
Positive test controls include: U-251MG, Rat brain, Rat thymus. Immunogen sequence: MNLAISIALL LTVLQVSRGQ KVTSLTACLV DQSLRLDCRH ENTSSSPIQY EFSLTRETKK HVLFGTVGVP EHTYRSRTNF TSKYNMKVLY LSAFTSKDEG.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.
Specifications
| CD90 (Thy1) | |
| Recombinant Monoclonal | |
| 0.53 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01830, P04216 | |
| Thy1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CD90/Thy1 (P04216). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2V9S6 | |
| Unconjugated | |
| Thy1 | |
| CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24832, 7070 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction