Learn More
CD90/Thy1 Antibody (CL1028) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52960AF647
Product Code. 30295388
  • £558.00 / 0.10mL
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD90/Thy1
Monoclonal
Alexa Fluor 647
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
This antibody was developed using a recombinant protein derived from P04216, with the exact immunogen sequence remaining proprietary.
0.1 mL
Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
7070
Store at 4⪚C in the dark.
IgG2b
Western Blot
CL1028
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified