missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Thy1 Antibody (AF-9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
£454.00 - £492.00
Specifications
| Antigen | CD90/Thy1 |
|---|---|
| Clone | AF-9 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606455
|
Novus Biologicals
NBP2-47755B-100ul |
100 μg |
£492.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692626
|
Novus Biologicals
NBP2-47755B |
0.1 mL |
£454.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| CD90/Thy1 | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 7070 | |
| IgG1 κ | |
| Protein A or G purified |
| AF-9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| Human T-lymphoma cells | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title