missing translation for 'onlineSavingsMsg'
Learn More

CD90/Thy1 Antibody (5E10), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18407922 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407922 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407922 Supplier Novus Biologicals Supplier No. NBP143379AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90/Thy1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5E10
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Host Species Mouse
Immunogen The immunogen for this antibody was CD90.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7070
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.