missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Mouse CD90.1 Mouse anti-Mouse, Rat, BUV563, Clone: OX-7 (also known as OX7), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 741304
This item is not returnable.
View return policy
Description
CD90/Mouse CD90.1 Monoclonal antibody specifically detects CD90/Mouse CD90.1 in Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| CD90/Mouse CD90.1 | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| Rat Thy-1; Mouse Thy-1.1 | |
| Rat Thymocyte Thy-1 Antigen | |
| RUO | |
| Mouse, Rat | |
| IgG1 κ |
| Flow Cytometry | |
| OX-7 (also known as OX7) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction