Learn More
Invitrogen™ CD90 Monoclonal Antibody (OX-7), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA180650
Description
This antibody recognizes rat and mouse CD90, also known as Thy1.1, expressed on a variety of cell types including thymocytes, neuronal cells, stem cells, T lymphocytes (mouse), immature B cells (rat) and connective tissues. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. Once reconstituted, this product has a shelf life of approx. 6 months. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.
Specifications
| CD90 | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.2 | |
| P01830, P01831 | |
| Thy1 | |
| Rat Thy1 antigen. | |
| 100 Tests | |
| Primary | |
| Mouse, Rat, Rabbit, Guinea Pig | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| OX-7 | |
| PE | |
| Thy1 | |
| CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
| Mouse | |
| Protein A | |
| RUO | |
| 100339279, 100379563, 21838, 24832 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.