Learn More
CD90.2 (Thy-1.2) Monoclonal Antibody (30-H12), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 13-0903-81
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The 30-H12 monoclonal antibody reacts with mouse CD90.2, also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/J, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 30-H12. CD90 is involved in regulation of adhesion and signal transduction by T cells. Applications Reported: The 30-H12 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 30-H12 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer′s cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.Specifications
CD90.2 (Thy-1.2) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P01831 | |
Thy1 | |
Affinity chromatography | |
RUO | |
21838 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
30-H12 | |
Biotin | |
Thy1 | |
CD90, T25, Thy-1, Thy-1.2, Thy1.1, Thy1.2,Thy 1.2; thy-1 antigen; thy-1 membrane glycoprotein | |
Rat | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
For Research Use Only.