missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90.2 Rat, PerCP-Cy5.5, Clone: 30-H12, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB52-PCP55-25ug
Additional Details : Weight : 0.01000kg
Specifications
CD90.2 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Thy1 | |
Protein A/G Purified | |
RUO | |
21838 | |
IgG2b |
Flow Cytometry | |
30-H12 | |
PerCP-Cyanine5.5 | |
NP_033408 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |