Learn More
CD90.2 Rat anti-Mouse, APC, Clone: 53-2.1, BD
Rat Monoclonal Antibody
£62.19 - £215.63
Specifications
| Antigen | CD90.2 |
|---|---|
| Clone | 53-2.1 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15891519
|
BD Biosciences
561974 |
25 μg |
£64.20 £62.19 / Each Save £2.01 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15896918
|
BD Biosciences
553007 |
0.1 mg |
£225.00 £215.63 / Each Save £9.37 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 53-2.1 monoclonal antibody specifically binds to the CD90.2 (Thy-1.2) alloantigen on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), epithelial cells, fibroblasts, neurons, hematopoietic stem cells, but not B lymphocytes, of most mouse strains. The 53-2.1 antibody has been reported not to crossreact with Thy-1.1 (e.g., AKR/J, PL), or with rat Thy-1. CD90 is a glycosyolphosphatidylinositol-anchored membrane glycoprotein of the Ig superfamily that is involved in signal transduction. In addition, there is evidence that CD90 mediates adhesion of thymocytes to thymic stroma. The 53-2.1 antibody has been reported to block the binding of anti-mouse CD90.2 clone 30-H12 (Cat. No. 553009) to immobilized thymocyte membranes.
Specifications
| CD90.2 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Mouse Thymus / Spleen | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 53-2.1 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| Thy-1.2; T25; Thymus cell antigen 1, theta |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.