Learn More
CD9 Mouse anti-Human, Unconjugated, Clone: MM2/57, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65070-1-IG-100UG
Description
CD9, also known as Tspan-29, p24 or MIC3, is a member of the tetraspanin superfamily (PMID: 1879540). It is expressed on a large variety of hematopoietic and non-hematopoietic cells, such as stromal cells, megakaryocytes, platelets, B and T lymphocytes, dendritic cells, endothelial cells, mast cells, eosinophils, and basophils (PMID: 30356731). CD9 interacts with the integrin family and other membrane proteins, and is involved in cell adhesion, cell motility and tumor metastasis (PMID: 8478605; 21428940; 25805926). Expression of CD9 enhances membrane fusion between muscle cells and support myotube maintenance (PMID:10459022). On oocytes, CD9 is hypothesized to play a role in fertilization of mammals (PMID: 25536312).
Specifications
| CD9 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Mouse | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| MM2/57 | |
| Unconjugated | |
| 100 μg | |
| 928 | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.