missing translation for 'onlineSavingsMsg'
Learn More

CD9 Mouse anti-Human, Clone: TS9, Novus Biologicals™

Product Code. 18389233
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389233 500 μg 500µL
18330107 200 μg 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18389233 Supplier Bio-Techne Supplier No. NBP314610500UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD9
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone TS9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunoprecipitation
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Host Species Mouse
Immunogen HeLa cell line
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 928
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.