missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD9 Monoclonal Antibody (MM2/57)

Mouse Monoclonal Antibody
Brand: Invitrogen™ AHS0902
This item is not returnable.
View return policy
Description
CD9 Monoclonal Antibody for Flow, IHC, IP, Western Blot
CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. CD9 belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41 / CD61 (GPIIb / GPIIIa). CD9 is involved in cell motility, osteoclastogenesis, neurite outgrowth, myotube formation, and sperm-egg fusion, plays roles in cell attachment and proliferation and is necessary for association of heterologous MHC II molecules on the dendritic cell plasma membrane which is important for effective T cell stimulation. CD9 functions in many cellular processes including differentiation, adhesion, and signal transduction, and expression plays a critical role in the suppression of cancer cell motility and metastasis. CD9 is also considered as metastasis suppressor in solid tumors.
Specifications
| CD9 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% BSA and 0.1% sodium azide | |
| P21926, P40240 | |
| CD9 | |
| Human platelet glycoprotein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rabbit | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| MM2/57 | |
| Unconjugated | |
| CD9 | |
| 5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100345844, 12527, 928 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction